Diacetyl 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Butanedione

10765-0050 10765-1000 10765-5000
TS10765-0050EA 56 USD
TS10765-0050 TS10765-1000 TS10765-5000
Diacetyl 99%
Diacetyl
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 4…2 °C
Density: 0.985 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008756
CAS Number: 431-03-8
EINECS: 207-069-8
UN: 2346
ADR: 3,II
Merck Index: 13,02985

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3990 (20 °C, 589 nm)
GC ≥98.5 %
Specific gravity (20°C) 0.98 to 0.99
Suitability (for Cu + Fe determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR