4-Phenylbenzaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Biphenyl-4-carbaldehyde, [1,1'-Biphenyl]-4-carbaldehyde, 4-Biphenylcarboxaldehyde, Biphenyl-4-carboxaldehyde, p-Biphenylylaldehyde, 4-Biphenylylaldehyde, 4-Biphenylcarbaldehyde
10626-0050
10626-0250
10626-1000
TS10626-0050EA
38
USD
TS10626-0050
TS10626-0250
TS10626-1000
4-Phenylbenzaldehyde 99%
4-Phenylbenzaldehyde
Formula:
C₁₃H₁₀O Boiling Pt: 184 °C (11 mmHg) Melting Pt: 57…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006947 CAS Number: 3218-36-8 EINECS: 221-742-3 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 57 to 60 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...