3-Benzoylpropionic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid

10579-0250 10579-1000
TS10579-0250EA 59 USD
TS10579-0250 TS10579-1000
3-Benzoylpropionic acid 98%
3-Benzoylpropionic acid
Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00002792
CAS Number: 2051-95-8
EINECS: 218-135-0

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 119 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR