9,10-Anthraquinone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Anthracene-9,10-dione, Anthraquinone

10493-0010 10493-0500 10493-2500
TS10493-0010EA 192 USD
TS10493-0010 TS10493-0500 TS10493-2500
9,10-Anthraquinone 98%
9,10-Anthraquinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 379…381 °C (760 mmHg)
Melting Pt: 283…288 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692

Order Now

Specification Test Results

Appearance Yellow-green to tan to khaki Powder
Infrared spectrum Conforms
Melting point 283 to 288 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR