Aminoguanidine nitrate 99%

Supplier: Thermo Scientific Chemicals
Warning

10376-0050 10376-0250 10376-1000
TS10376-0050EA 18 USD
TS10376-0050 TS10376-0250 TS10376-1000
Aminoguanidine nitrate 99%
Aminoguanidine nitrate
Formula: CH₇N₅O₃
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00013174
CAS Number: 10308-82-4
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 147 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR