2-(2-Aminoethoxy)ethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

10363-0010 10363-0025 10363-1000
TS10363-0010EA 157 USD
TS10363-0010 TS10363-0025 TS10363-1000
2-(2-Aminoethoxy)ethanol 98%
2-(2-Aminoethoxy)ethanol
Formula: C₄H₁₁NO₂
Boiling Pt: 218…224 °C (760 mmHg)
Melting Pt: –12.5 °C
Density: 1.06 g/cm³
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008181
CAS Number: 929-06-6
EINECS: 213-195-4
UN: 3055
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4610 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR