4'-Aminoacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

10309-0100 10309-0250 10309-1000 10309-5000
TS10309-0100EA 24 USD
TS10309-0100 TS10309-0250 TS10309-1000 TS10309-5000
4'-Aminoacetophenone 99%
4'-Aminoacetophenone
Formula: C₈H₉NO
Boiling Pt: 293 °C (760 mmHg)
Melting Pt: 96…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00007896
CAS Number: 99-92-3
EINECS: 202-801-2
Merck Index: 13,00412

Order Now

Specification Test Results

Appearance Light yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 104 to 107 °C
GC ≥98.5 %
Absorption Ratio P-15/P+15 1.16 to 1.35
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR