Methyl hydrogen adipate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,6-Hexanedioic acid methyl ester, Adipic acid methyl ester, Adipic acid monomethyl ester, 1,6-Hexanedioic acid monomethyl ester, Monomethyl adipate

10282-0050 10282-0250 10282-1000
TS10282-0050EA 35 USD
TS10282-0050 TS10282-0250 TS10282-1000
Methyl hydrogen adipate 97%
Methyl hydrogen adipate
Formula: C₇H₁₂O₄
Boiling Pt: 162 °C (10 mmHg)
Melting Pt: 8…9 °C
Density: 1.08 g/cm³
Flash Pt: >109 °C
MDL Number: MFCD00004418
CAS Number: 627-91-8
EINECS: 211-019-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20 °C, 589 nm)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR