Acrolein diethyl acetal 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene
10277-0250
10277-1000
TS10277-0250EA
77
USD
TS10277-0250
TS10277-1000
Acrolein diethyl acetal 94%
Acrolein diethyl acetal
Formula:
C₇H₁₄O₂ Boiling Pt: 120…125 °C (760 mmHg) Melting Pt: –77 °C Density: 0.85 g/cm³ Flash Pt: 15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009239 CAS Number: 3054-95-3 UN: 2374 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3970 to 1.4030 (20 °C, 589 nm) |
GC | ≥92 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (ICP-OES) | ≤5 ppm |
Sulfur compounds (as SO4) | ≤0.002 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...