4-Acetylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-pyridyl ketone

10270-0100 10270-1000 10270-5000
TS10270-0100EA 26 USD
TS10270-0100 TS10270-1000 TS10270-5000
4-Acetylpyridine 98%
4-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 212 °C (760 mmHg)
Melting Pt: 13…16 °C
Density: 1.09 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00006433
CAS Number: 1122-54-9
EINECS: 214-350-9

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5270 to 1.5310 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR