3-Acetylpyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone

10269-0100 10269-1000 10269-5000
TS10269-0100EA 31 USD
TS10269-0100 TS10269-1000 TS10269-5000
3-Acetylpyridine 98%
3-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 12…13 °C
Density: 1.1 g/cm³
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1,III
Merck Index: 12,06195

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR