2-Acetylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10268-0250
10268-1000
10268-5000
TS10268-0250EA
49
USD
TS10268-0250
TS10268-1000
TS10268-5000
2-Acetylpyridine 98%
2-Acetylpyridine
Formula:
C₇H₇NO Boiling Pt: 188…189 °C (760 mmHg) Melting Pt: 8…10 °C Density: 1.08 g/cm³ Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006303 CAS Number: 1122-62-9 EINECS: 214-355-6 |
Specification Test Results
Appearance | Clear colorless to yellow to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5250 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...