3-Acetyl-2,4-dimethylpyrrole 98%

Supplier: Thermo Scientific Chemicals
Warning

10251-0100 10251-0500
TS10251-0100EA 444 USD
TS10251-0100 TS10251-0500
3-Acetyl-2,4-dimethylpyrrole 98%
3-Acetyl-2,4-dimethylpyrrole
Formula: C₈H₁₁NO
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00005221
CAS Number: 2386-25-6

Order Now

Specification Test Results

Appearance Light yellow to pink to brown to purple Powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR