2-Acetylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

10247-0050 10247-0250 10247-1000
TS10247-0050EA 44 USD
TS10247-0050 TS10247-0250 TS10247-1000
2-Acetylbenzoic acid 99%
2-Acetylbenzoic acid
Formula: C₉H₈O₃
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002475
CAS Number: 577-56-0
EINECS: 209-413-2

Order Now

Specification Test Results

Appearance Light yellow to light orange Crystalline powder
Infrared spectrum Conforms
Melting point 115 to 118 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR