4'-Hydroxy-3'-methoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

10242-0250 10242-1000 10242-5000
TS10242-0250EA 35 USD
TS10242-0250 TS10242-1000 TS10242-5000
4'-Hydroxy-3'-methoxyacetophenone 98%
4'-Hydroxy-3'-methoxyacetophenone
Formula: C₉H₁₀O₃
Boiling Pt: 295…300 °C (760 mmHg)
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00008747
CAS Number: 498-02-2
EINECS: 207-854-5
Merck Index: 13,00746

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
Water ≤0.5 % (K.F.)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR