Vornado Mini Vortex Mixers, Southern Labware
Supplier: SOUTHERN LABWARE, INC MS
BV101-R BV101-B
MSPP-BV101-REA 293.71 USD
MSPP-BV101-R MSPP-BV101-B
Vornado Mini Vortex Mixers, Southern Labware
Shakers and Mixers Vortex Mixers
The Vornado mini vortexer has a 4 mm orbit and fixed speed of 2800 rpm instantly mixing even the largest samples, including nearly full 50 ml tubes. Despite its powerful motor, the Vornado has a footprint of less than 4×4", allowing it to fit on even the most crowded bench.
  • Powerful vortexing for tubes up to 50 ml
  • Inst-Touch™ pressure activated operation
  • 'Mini Sized', less than 4" wide
  • Available in different color configutaions
  • Two years warranty
Insta-Touch operation means that the motor is automatically activated with slight pressure on the centerMore Product Information