Dipropylene glycol monobutyl ether

Supplier: Thermo Scientific Chemicals

Synonyms: ppg-2 butyl ether, 1-[(1-butoxypropan-2-yl)oxy]propan-2-ol

46726-0010 46726-0025 46726-0100
TS46726-0010EA 43 USD
TS46726-0010 TS46726-0025 TS46726-0100
Dipropylene glycol monobutyl ether
Dipropylene glycol monobutyl ether
Formula: C₁₀H₂₂O₃
MW: 190.28 g/mol
Boiling Pt: 230 °C
Density: 0.91
CAS Number: 29911-28-2
EINECS: 249-951-5

Order Now

Specification Test Results

Appearance (Color) Clear colorless
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, nm)
Appearance (Form) Liquid
GC >=97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR