Triethylene glycol dimethacrylate contains MEHQ as inhibitor

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(2-methylacrylate)

46627-0010 46627-2500
TS46627-0010EA 237 USD
TS46627-0010 TS46627-2500
Triethylene glycol dimethacrylate contains MEHQ as inhibitor
Triethylene glycol dimethacrylate

Viscosity 11 mPa.s (25°C)

Formula: C₁₄H₂₂O₆
MW: 286.32 g/mol
Boiling Pt: 170.0°C - 172.0°C (5.0 mmHg)
Density: 1.0920 g/mL
Flash Pt: 167 °C
MDL Number: MFCD00008591
CAS Number: 109-16-0
EINECS: 203-652-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR