Dypnone

Supplier: Thermo Scientific Chemicals

46585-0050 465850250
TS46585-0050EA 127 USD
TS46585-0050 TS465850250
Dypnone
Dypnone
Formula: C₁₆H₁₄O
MDL Number: MFCD00043705
CAS Number: 495-45-4

Order Now

Specification Test Results

Appearance Colorless to yellow liquid
Infrared Spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR