Oil for heat transfer

Supplier: Thermo Scientific Chemicals

26877-0025 26877-0100
TS26877-0025EA 100 USD
TS26877-0025 TS26877-0100
Oil for heat transfer
Oil
CAS Number: 64742-65-0
EINECS: 265-169-7

Order Now

Specification Test Results

Appearance (Form) Oily liquid
Viscosity 28.0 to 32.0 cSt (at 40°C) (kinematic)
Density (15°C) 0.84 to 0.88 kg/l
Appearance (Color) Clear light yellow
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR