Divosan® Plus, Non-foaming Liquid Peroxyacetic Acid Sanitizer
Supplier: DIVERSEY INC.
3177186 3747804 57571100
76547-690EA 1728.62 USD
76547-690 76547-692 76547-730
Divosan® Plus, Non-foaming Liquid Peroxyacetic Acid Sanitizer
Cleaning Agents Detergents and Surface Cleansers
Divosan® Plus is a peracetic acid based, broad spectrum sanitizer.
  • Completely non-foaming and suitable for clean-in-place (CIP) and spray applications
  • Rapidly decomposes after use
  • Sanitizes (reduces bacteria) in 1 minute, controls spoilage organisms; controlsmolds and yeast
  • Effective at temperatures as low as 40°F
  • Acidic pH, suitable under CO2 environments
  • 1.0 - 5.7 ounces per 5 gallons of water (88-500 ppm peracetic acid)
It is suitable for use in dairies, breweries, soft drink and all other food processing operations.
Divosan Plus is completely non-foaming and sanitizes effectively at temperatures as low as 40°F.