2,8-Bis(trifluoromethyl)quinolin-4-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

30601-0010 306010050
TS30601-0010EA 21 USD
TS30601-0010 TS306010050
2,8-Bis(trifluoromethyl)quinolin-4-ol 99%
2,8-Bis(trifluoromethyl)quinolin-4-ol
Formula: C₁₁H₅F₆NO
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00075091
CAS Number: 35853-41-9

Order Now

Specification Test Results

Appearance White to brown-yellow to light grey Powder
Infrared spectrum Conforms
Melting point 128 to 134 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR