2,2,2-Trifluorothanesulphonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,2-Trifluoroethanesulphonyl chloride, (2,2,2-Trifluoroethyl)sulphonyl chloride

30535-0100 30535-0010 30535-0025
TS30535-0100EA 676 USD
TS30535-0100 TS30535-0010 TS30535-0025
2,2,2-Trifluorothanesulphonyl chloride 99%
2,2,2-Trifluorothanesulphonyl chloride
Formula: C₂H₂ClF₃O₂S
MW: 182.55 g/mol
Boiling Pt: 64…67 °C (45 mmHg)
Density: 1.65 g/cm³
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007458
CAS Number: 1648-99-3
EINECS: 216-713-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3890 (20°C, 589 nm)
Titration Argentometric ≥98.5 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR