2-Ethylhexyl bromide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane

30533-1000 30533-5000
TS30533-1000EA 44 USD
TS30533-1000 TS30533-5000
2-Ethylhexyl bromide 95%
2-Ethylhexyl bromide
Formula: C₈H₁₇Br
MW: 193.13 g/mol
Boiling Pt: 75…77 °C (16 mmHg)
Density: 1.08 g/cm³
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000220
CAS Number: 18908-66-2
EINECS: 242-659-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4560 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR