2-Iodo-1-phenylethane 97% stabilized

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Phenylethyl iodide, (2-Iodoethyl)benzene

Stabilization: Stabilised with copper

30525-0050 30525-0250
TS30525-0050EA 83 USD
TS30525-0050 TS30525-0250
2-Iodo-1-phenylethane 97% stabilized
2-Iodo-1-phenylethane
Formula: C₆H₅CH₂CH₂I
MW: 232.06 g/mol
Boiling Pt: 122…123 °C (13 mmHg)
Melting Pt: –2 °C
Density: 1.6 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00039408
CAS Number: 17376-04-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5990 to 1.6030 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR