2,2,3,3,3-Pentafluoro-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H,1H-Pentafluoro-1-propanol, PFPOH (pentafluoropropanol), 1H,1H-Pentafluoropropanol-1, 2,2,3,3,3-Pentafluoropropanol

30379-0050 30379-0250
TS30379-0050EA 39 USD
TS30379-0050 TS30379-0250
2,2,3,3,3-Pentafluoro-1-propanol 97%
2,2,3,3,3-Pentafluoro-1-propanol
Formula: C₃H₃F₅O
Boiling Pt: 78…80 °C (1013 hPa)
MDL Number: MFCD00004673
CAS Number: 422-05-9
EINECS: 207-012-7
UN: 1986

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR