Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
30213-0050
30213-0250
TS30213-0050EA
135
USD
TS30213-0050
TS30213-0250
Bromotrispyrrolidinophosphonium hexafluorophosphate 97%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Formula:
C₁₂H₂₄BrF₆N₃P₂ MW: 466.2 g/mol Melting Pt: >100 °C Storage Temperature: Freezer |
MDL Number:
MFCD00077412 CAS Number: 132705-51-2 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | ≥100 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...