4-Hydroxy-7-methoxycoumarin 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-7-methoxy-2H-chromen-2-one

29885-0010 29885-0050
TS29885-0010EA 85 USD
TS29885-0010 TS29885-0050
4-Hydroxy-7-methoxycoumarin 95%
4-Hydroxy-7-methoxycoumarin
Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: 263…265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00673700
CAS Number: 17575-15-4

Order Now

Specification Test Results

Appearance Beige to pink Powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR