2,2,3,3-Tetrafluoro-1-propanol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,3,3-Tetrafluoropropan-1-ol, 1H,1H,3H-Tetrafluoro-1-propanol

29638-0250 29638-1000
TS29638-0250EA 77 USD
TS29638-0250 TS29638-1000
2,2,3,3-Tetrafluoro-1-propanol 99+%
2,2,3,3-Tetrafluoro-1-propanol
Formula: C₃H₄F₄O
MW: 132.06 g/mol
Boiling Pt: 109 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.47 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00004676
CAS Number: 76-37-9
EINECS: 200-955-5
UN: 1986
ADR: 3,III
Merck Index: 12,09346

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3200 to 1.3220 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR