Chloromethyl butyrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloromethylbutyrate, Butyric Acid Chloromethyl Ester

29558-0050 29558-0250
TS29558-0050EA 66 USD
TS29558-0050 TS29558-0250
Chloromethyl butyrate 99%
Chloromethyl butyrate
Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 150 °C (993mbar)
Density: 1.074 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00216961
CAS Number: 33657-49-7
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR