Sebacoyl dichloride 92%, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sebacoyl chloride

29490-2500 29490-0500 29490-5000
TS29490-2500EA 165 USD
TS29490-2500 TS29490-0500 TS29490-5000
Sebacoyl dichloride 92%, Technical Grade
Sebacoyl dichloride
Formula: C₁₀H₁₆Cl₂O₂
MW: 239.14 g/mol
Boiling Pt: 220 °C (75 mmHg)
Melting Pt: –2.5 °C
Density: 1.121 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000770
CAS Number: 111-19-3
EINECS: 203-843-4
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to golden to brown Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4700 (20°C, 589 nm)
GC ≥90 %
Titration RCOCl ≥90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR