diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide

27781-0010 27781-2500
TS27781-0010EA 527 USD
TS27781-0010 TS27781-2500
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide 99+%
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
Formula: C₈H₁₂N₂O
MW: 152.197 g/mol
Melting Pt: 235…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00143157
CAS Number: 105786-40-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR