1H,1H-Pentadecafluoro-1-octanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H,1H-Perfluoro-1-octanol, 1H,1h-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluor-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octanol

27726-0050 27726-0250
TS27726-0050EA 56 USD
TS27726-0050 TS27726-0250
1H,1H-Pentadecafluoro-1-octanol 98%
1H,1H-Pentadecafluoro-1-octanol
Formula: C₈H₃F₁₅O
MW: 400.09 g/mol
Boiling Pt: 100…102 °C (100hPa)
Melting Pt: 43…47 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004675
CAS Number: 307-30-2
EINECS: 206-197-1

Order Now

Specification Test Results

Appearance White Crystalline mass
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR