1-Naphthoyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

27038-0100 27038-0500
TS27038-0100EA 56 USD
TS27038-0100 TS27038-0500
1-Naphthoyl chloride 99%
1-Naphthoyl chloride
Formula: C₁₁H₇ClO
MW: 190.63 g/mol
Boiling Pt: 190 °C (35 mmHg)
Melting Pt: 16…19 °C
Density: 1.26 g/cm³
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00004002
CAS Number: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear yellow to beige to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6510 to 1.6540 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR