1-Iodoperfluorooctane 98%

Supplier: Thermo Scientific Chemicals
Warning

26996-0100 26996-0500
TS26996-0100EA 38 USD
TS26996-0100 TS26996-0500
1-Iodoperfluorooctane 98%
1-Iodoperfluorooctane
Formula: C₈F₁₇I
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: 25 °C
Density: 2.04 g/cm³
Flash Pt: >115 °C
Storage Temperature: Ambient
MDL Number: MFCD00001064
CAS Number: 507-63-1
EINECS: 208-079-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light pink Liquid after melting
Infrared spectrum Conforms
Refractive index 1.3280 to 1.3310 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR