1-Iodoperfluorbutane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane

26995-0250 26995-1000
TS26995-0250EA 61 USD
TS26995-0250 TS26995-1000
1-Iodoperfluorbutane 99%
1-Iodoperfluorbutane
Formula: C₄F₉I
MW: 345.93 g/mol
Boiling Pt: 64…66 °C (973 mbar)
Melting Pt: –88 °C
Density: 2.01 g/cm³
Flash Pt: >115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001062
CAS Number: 423-39-2
EINECS: 207-025-8

Order Now

Specification Test Results

Appearance Clear purple Liquid
Infrared spectrum Conforms
Refractive index 1.3275 to 1.3295 (20°C, 589 nm)
GC ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR