Triisopropanolamine cyclic borate 98+%

Supplier: Thermo Scientific Chemicals

26924-1000 26924-5000
TS26924-1000EA 205 USD
TS26924-1000 TS26924-5000
Triisopropanolamine cyclic borate 98+%
Triisopropanolamine cyclic borate
Formula: C₉H₁₈BNO₃
Melting Pt: 150…157 °C
Storage Temperature: Ambient
MDL Number: MFCD03791298
CAS Number: 101-00-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline needles and/or chunks
Infrared spectrum Conforms
Melting point 150 to 157 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR