2-(Methyldithio)isobutyraldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

26515-0010 26515-0050
TS26515-0010EA 159 USD
TS26515-0010 TS26515-0050
2-(Methyldithio)isobutyraldehyde 95%
2-(Methyldithio)isobutyraldehyde
Formula: C₅H₁₀OS₂
Boiling Pt: 46 °C (1 mmHg)
Density: 1.1 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00142943
CAS Number: 67952-60-7

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5230 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR