4-Amino-6-chlorobenzene-1,3-disulphonamide 98%
Supplier: Thermo Scientific Chemicals
25185-0050
25185-1000
TS25185-0050EA
27
USD
TS25185-0050
TS25185-1000
4-Amino-6-chlorobenzene-1,3-disulphonamide 98%
4-Amino-6-chlorobenzene-1,3-disulphonamide
Formula:
C₆H₈ClN₃O₄S₂ Melting Pt: 260…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007933 CAS Number: 121-30-2 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 260 to 264 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...