(±)-3-Butyne-2-ol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (±)-But-3-yn-2-ol, (±)-3-Hydroxy-1-butyne

24531-0100 24531-0500
TS24531-0100EA 44 USD
TS24531-0100 TS24531-0500
(±)-3-Butyne-2-ol 97%
(±)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 104…108 °C (1013 hPa)
Melting Pt: -12
Density: 0.89 g/cm³
Flash Pt: 26 °C
MDL Number: MFCD00004541
CAS Number: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4270 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR