2,4,6-Trimethylbenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4,6-Trimethylbenzene-1-carbonyl chloride, 2,4,6-Trimethylbenzoyl Chloride

24428-0100 24428-0500
TS24428-0100EA 180 USD
TS24428-0100 TS24428-0500
2,4,6-Trimethylbenzoyl chloride 98%
2,4,6-Trimethylbenzoyl chloride
Formula: C₁₀H₁₁ClO
MW: 182.65 g/mol
Boiling Pt: 143…146 °C (60 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00013650
CAS Number: 938-18-1
EINECS: 213-339-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5280 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR