2,2,3,3,4,4,4-Heptafluoro-1-butanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol

24389-0100 24389-0500
TS24389-0100EA 59 USD
TS24389-0100 TS24389-0500
2,2,3,3,4,4,4-Heptafluoro-1-butanol 95%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 96…97 °C (760 mmHg)
Density: 1.6 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00004674
CAS Number: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR