1-Octyne 99%

Supplier: Thermo Scientific Chemicals
Danger

22351-0250 22351-1000
TS22351-0250EA 69 USD
TS22351-0250 TS22351-1000
1-Octyne 99%
1-Octyne
Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 127…128 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.74 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00009546
CAS Number: 629-05-0
EINECS: 211-069-3
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR