(Bromomethyl)cyclopropane 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
22181-0250
22181-0010
22181-0050
TS22181-0250EA
385
USD
TS22181-0250
TS22181-0010
TS22181-0050
(Bromomethyl)cyclopropane 96%
(Bromomethyl)cyclopropane
Formula:
C₄H₇Br MW: 135 g/mol Boiling Pt: 101…108 °C (760 mmHg) Density: 1.38 g/cm³ Flash Pt: 41 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001306 CAS Number: 7051-34-5 EINECS: 230-331-8 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to slightly colored Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4750 to 1.4790 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥95 % |
Specific gravity | (20°C) 1.385 to 1.415 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...