1-Octanol 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Octanol

22018-0010 22018-0025
TS22018-0010EA 92 USD
TS22018-0010 TS22018-0025
1-Octanol 99+% ACS
1-Octanol
Formula: H₃C(CH₂)₆CH₂OH
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –16 °C
Density: 0.8246 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784

Order Now

Specification Test Results

Appearance (Color) Clear colorless
Appearance (Form) Liquid
Color scale =<10 APHA
GC >=99 %
Residue after evaporation =<0.004 %
Titrable acid =<0.0002 meq/g
Refractive index 1.4280 to 1.4310 (20°C, 589nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR