1,3-Diiodopropane 99% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylene diiodide

21860-0250 21860-1000
TS21860-0250EA 115 USD
TS21860-0250 TS21860-1000
1,3-Diiodopropane 99% stabilized
1,3-Diiodopropane
Formula: C₃H₆I₂
MW: 295.89 g/mol
Boiling Pt: 111…113 °C (31 mmHg)
Melting Pt: –20 °C
Density: 2.576 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00001097
CAS Number: 627-31-6
EINECS: 210-993-4

Order Now

Specification Test Results

Appearance Clear light yellow to pink Liquid
Infrared spectrum Conforms
Refractive index 1.6405 to 1.6425 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR