3-Chloro-1-iodopropane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-3-iodopropane

21544-0250 21544-1000
TS21544-0250EA 64 USD
TS21544-0250 TS21544-1000
3-Chloro-1-iodopropane 98% stabilized
3-Chloro-1-iodopropane
Formula: C₃H₆ClI
MW: 204.44 g/mol
Boiling Pt: 171 °C (1013 hPa)
Density: 1.9 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001096
CAS Number: 6940-76-7
EINECS: 230-088-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5440 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR