1-Bromohexadecane 97%

Supplier: Thermo Scientific Chemicals

21543-0010 21543-0025 21543-2500
TS21543-0010EA 178 USD
TS21543-0010 TS21543-0025 TS21543-2500
1-Bromohexadecane 97%
1-Bromohexadecane
Formula: BrH₂C(CH₂)₁₄CH₃
MW: 305.34 g/mol
Boiling Pt: 336 °C (760 mmHg)
Melting Pt: 17.3 °C
Density: 0.999 g/cm³
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00000230
CAS Number: 112-82-3
EINECS: 204-008-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow to amber to brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4640 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR