Phenyl phosphate, disodium salt dihydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium phenyl phosphate dibasic dihydrate

21183-0100 21183-1000
TS21183-0100EA 56 USD
TS21183-0100 TS21183-1000
Phenyl phosphate, disodium salt dihydrate 98%
Phenyl phosphate, disodium salt dihydrate
Formula: C₆H₉Na₂O₆P
MW: 254.085 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149080
CAS Number: 66778-08-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water 12 to 16 % (K.F.)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR