Acriflavine neutral 13.3-15.8% available chlorine

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base

21140-0250 21140-1000
TS21140-0250EA 71 USD
TS21140-0250 TS21140-1000
Acriflavine neutral 13.3-15.8% available chlorine
Acriflavine
Formula: C₂₇H₂₅ClN₆
MW: 468.9806 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00064307
CAS Number: 8048-52-0

Order Now

Specification Test Results

Appearance Orange to red-brown Powder
Infrared spectrum Conforms
Titration Argentometric 13.3 to 15.8 % (Chlorine) on dried substance
Loss on drying ≤8 % (105°C, 2 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR